Learn More
Macherey-Nagel Syringe filters CHROMAFIL PET-45/25
Supplier:  Macherey-Nagel 729020
Encompass_Preferred
CHROMAFIL disposable syringe filters PET-45/25 - membrane material: polyester - pore dia.: 0.45 µm, membrane dia.: 25 mm - color code above: colorless / below: orange - pack of 100
Catalog No. NC1227175