Learn More
Epigentek Inc ChromaFlash Chromatin Isolation & Shearing Kit (100 preparations)
SDP
Catalog No. 501096168
Encompass_Preferred
The ChromaFlash Chromatin Isolation & Shearing Kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells or tissues in a simple and rapid format, followed by a shearing step by sonication. Chromatin prepared by this kit can be used in a variety of chromatin immunoprecipitation methods. It is also the recommended method for obtaining chromatin required by Epigentek’s one-hour ChIP method using the ChromaFlash One-Step ChIP Kit. The isolated chromatin can also be used in other chromatin-related applications such as in vitro protein-DNA binding assays and nuclear enzyme assays.
Catalog No. 50-109-6168 Supplier Epigentek Inc Supplier No. P2023100
The ChromaFlash Chromatin Isolation & Shearing Kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells or tissues in a simple and rapid format, followed by a shearing step by sonication. Chromatin prepared by this kit can be used in a variety of chromatin immunoprecipitation methods. It is also the recommended method for obtaining chromatin required by Epigentek’s one-hour ChIP method using the ChromaFlash One-Step ChIP Kit. The isolated chromatin can also be used in other chromatin-related applications such as in vitro protein-DNA binding assays and nuclear enzyme assays.