Learn More
Vector Laboratories ChromaLINK Biotin Protein Kit B-9007-009K
SDP
Catalog No. 502045723
Encompass
ChromaLINK Biotin Protein Labeling Kit, 1 Kit, includes everything you need to biotinylate your protein: buffers, reagents, spin columns, and an easy-to-follow protocol and online calculator. With the ChromaLINK Biotin Protein Labeling Kit, you can biotinylate and purify up to 5 antibodies or other proteins ranging in molecular weight from 20–200 kDa and from as little as 25 µg to as much as 1 mg.
Catalog No. 50-204-5723 Supplier Vector Laboratories Supplier No. B9007009K
ChromaLINK Biotin Protein Labeling Kit, 1 Kit, includes everything you need to biotinylate your protein: buffers, reagents, spin columns, and an easy-to-follow protocol and online calculator. With the ChromaLINK Biotin Protein Labeling Kit, you can biotinylate and purify up to 5 antibodies or other proteins ranging in molecular weight from 20–200 kDa and from as little as 25 µg to as much as 1 mg.