Learn More
Vector Laboratories ChromaLINK Biotin (DMF Soluble), 5 x 1 mg
SDP
Catalog No. 502045719
Encompass
Bioconjugation. ChromaLINK Biotin contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches.
Catalog No. 50-204-5719 Supplier Vector Laboratories Supplier No. B1001105
Bioconjugation. ChromaLINK Biotin contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches.