Learn More
Vector Laboratories ChromaLINK Biotin Maleimide B-9007-105K
SDP
Catalog No. 502045724
Encompass
ChromaLINK Biotin Maleimide, 10 mg, contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches. The maleimide group reacts with free thiol groups on biomolecules and surfaces, such as the hinge region of antibodies after mild reduction of sulfhydryl groups with TCEP or DTT.
Catalog No. 50-204-5724 Supplier Vector Laboratories Supplier No. B9007105K
ChromaLINK Biotin Maleimide, 10 mg, contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches. The maleimide group reacts with free thiol groups on biomolecules and surfaces, such as the hinge region of antibodies after mild reduction of sulfhydryl groups with TCEP or DTT.