Learn More
Vector Laboratories ChromaLINK Biotin, 5 x 1 mg B-1012-010
SDP
Supplier:  Vector Laboratories B1012010
Encompass
ChromaLINK Biotin (DMF Soluble), 5 x 1 mg, contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches.
Catalog No. 50-204-5722