Learn More
Medchemexpress LLC HY-15392 5mg Medchemexpress, Chroman 1 CAS:1273579-40-0 Purity:>98%
SDP
Catalog No. 502029121
Encompass_Preferred
Medchemexpress, HY-15392 5mg Chroman 1 CAS:1273579-40-0 Chroman 1 dihydrochloride is a highly potent ROCK2 inhibitor, with an IC50 of 1 nM. Chroman 1 dihydrochloride also shows inhibitory activities against MRCK, with an IC50 of 150 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9121 Supplier Medchemexpress LLC Supplier No. HY153925MG
Medchemexpress, HY-15392 5mg Chroman 1 CAS:1273579-40-0 Chroman 1 dihydrochloride is a highly potent ROCK2 inhibitor, with an IC50 of 1 nM. Chroman 1 dihydrochloride also shows inhibitory activities against MRCK, with an IC50 of 150 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.