Learn More
Medchemexpress LLC Chroman 1 dihydrochloride | 1273579-40-0 | C24H30Cl2N4O4 | 25 MG
SDP
Catalog No. 5000375072
Encompass_Preferred
Chroman 1 dihydrochloride is a highly potent and selective ROCK inhibitor, demonstrating greater potency against ROCK2 (IC50=1 pM) compared to ROCK1 (IC50=52 pM). It also exhibits inhibitory activity against MRCK (IC50=150 nM) and has been shown to inhibit caspase-3/7 activation and reduce apoptosis in human pluripotent stem cells.
  • Highly potent and selective ROCK inhibitor
  • More potent against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM)
  • Exhibits inhibitory activity against MRCK (IC50=150 nM)
  • Inhibits caspase-3/7 activation
  • Reduces apoptosis in human pluripotent stem cells
Catalog No. 50-003-75072 Supplier Medchemexpress LLC Supplier No. 25MGHY15392A
Chroman 1 dihydrochloride is a highly potent and selective ROCK inhibitor, demonstrating greater potency against ROCK2 (IC50=1 pM) compared to ROCK1 (IC50=52 pM). It also exhibits inhibitory activity against MRCK (IC50=150 nM) and has been shown to inhibit caspase-3/7 activation and reduce apoptosis in human pluripotent stem cells.
  • Highly potent and selective ROCK inhibitor
  • More potent against ROCK2 (IC50=1 pM) than ROCK1 (IC50=52 pM)
  • Exhibits inhibitory activity against MRCK (IC50=150 nM)
  • Inhibits caspase-3/7 activation
  • Reduces apoptosis in human pluripotent stem cells