Learn More
Medchemexpress LLC Chroman 1 dihydrochloride | C24H30Cl2N4O4 | 100 MG
SDP
Catalog No. 5000376110
Encompass_Preferred
Chroman 1 dihydrochloride is a highly potent and selective ROCK inhibitor. It demonstrates greater potency against ROCK2 than ROCK1, and also inhibits MRCK. This compound has been shown to reduce apoptosis and caspase-3/7 activation in human pluripotent stem cells.
  • Highly potent and selective ROCK inhibitor
  • More potent against ROCK2 (IC50=1 pM)
  • Inhibits ROCK1 (IC50=52 pM)
  • Inhibits MRCK (IC50=150 nM)
  • Reduces apoptosis in human pluripotent stem cells
  • Inhibits caspase-3/7 activation in human pluripotent stem cells
Catalog No. 50-003-76110 Supplier Medchemexpress LLC Supplier No. HY15392A100MG
Chroman 1 dihydrochloride is a highly potent and selective ROCK inhibitor. It demonstrates greater potency against ROCK2 than ROCK1, and also inhibits MRCK. This compound has been shown to reduce apoptosis and caspase-3/7 activation in human pluripotent stem cells.
  • Highly potent and selective ROCK inhibitor
  • More potent against ROCK2 (IC50=1 pM)
  • Inhibits ROCK1 (IC50=52 pM)
  • Inhibits MRCK (IC50=150 nM)
  • Reduces apoptosis in human pluripotent stem cells
  • Inhibits caspase-3/7 activation in human pluripotent stem cells