Learn More
Selleck Chemical LLC Chroman 1-E1261-5MG
SDP
Catalog No. 503137250
Encompass_Preferred
Chroman 1 is a highly potent and selective ROCK inhibitor with IC50 of 1 pM and 52 pM against ROCK2 and ROCK1 respectively Chroman 1 also inhibits the activity of MRCK with IC50 of 150 nM
Catalog No. 50-313-7250 Supplier Selleck Chemical LLC Supplier No. E12615MG
Add to Cart
Add to Cart
Chroman 1 is a highly potent and selective ROCK inhibitor with IC50 of 1 pM and 52 pM against ROCK2 and ROCK1 respectively Chroman 1 also inhibits the activity of MRCK with IC50 of 150 nM