Learn More
Medchemexpress LLC HY-15392 10mg , Chroman 1 ROCK-II inhibitor CAS:1273579-40-0 Purity:98%
SDP
Catalog No. 501494064
Encompass_Preferred
Medchemexpress, HY-15392 10mg Chroman 1 ROCK-II inhibitor CAS:1273579-40-0 Formula:C24H28N4O4 IC50: < 1 nM (ROCK2) Purity:98% Chroman 1 is a highly potent ROCK2 inhibitor, with an IC 50 of < 1 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4064 Supplier Medchemexpress LLC Supplier No. HY1539210MG
Medchemexpress, HY-15392 10mg Chroman 1 ROCK-II inhibitor CAS:1273579-40-0 Formula:C24H28N4O4 IC50: < 1 nM (ROCK2) Purity:98% Chroman 1 is a highly potent ROCK2 inhibitor, with an IC 50 of < 1 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.