Learn More
Synthware™ Chromaography Reservior with Rodaviss Joint
GREENER_CHOICESDP
Catalog No. 31501005 Shop All Kemtech Products
Change view
Click to view available options
Description:
Joint: 24/50; 250 mL
Joint: 24/50; 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
31-501-005 Joint: 24/50; 250 mL
31-501-006 Joint: 24/50; 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-005 Supplier Synthware™ Supplier No. C414250
Add to Cart
Add to Cart
Designed for use with flash chromatography. Supplied complete with open top cap, Viton O-ring and loosening ring.
Specifications
Technique Chromatography
Quantity 1 Ea.
Description Joint: 24/50; 250 mL
Material Borosilicate Glass