Learn More
CHEMetrics Chromate CHEMets Refill



Specifications
Specifications
| Analytes | Chromate, Hexavalent |
| Format | Liquid |
| For Use With (Equipment) | C-2801 and C-2810 comparators |
| Includes | 30 self-filling CHEMets ampoules to refill kit K-2810 |
| No. per Pack | 30 |
| Quantity | 30 ampoules |
| Test Method | Diphenylcarbazide |
| Type | Chromate Refill |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.