Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChromaTide™ Alexa Fluor™ 568-5-dUTP

Catalog No. C11399
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11399 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11399 Supplier Invitrogen™ Supplier No. C11399
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ChromaTide™ Alexa Fluor™ 568-5-dUTP

Molecular Probes™ ChromaTide™ dye labeled dUTP, OBEA-dCTP, and UTP nucleotides can be used to synthesize labeled DNA probes without the need for hazardous and expensive radioisotope-labeled nucleotides. These nucleotides can be incorporated using standard molecular biology techniques; labeled probes can then be used in in situ hybridization, microarray, or blotting protocols. ChromaTide™ dye labeled nucleotides are available in different fluorescent colors to facilitate multicolor analysis.

ChromaTide™ Labeled Nucleotides Specifications:
  • Ex/Em of dye: Alexa Fluor™ 568-5-dUTP (575/600 nm)
  • Length of alkynylamino linker: 5 atoms

    Methods for Incorporating ChromaTide™ Nucleotides Into Probes
  • Nick translation
  • Random primer labeling
  • End-labeling with terminal deoxynucleotidyl transferase
  • Reverse transcription
  • PCR amplification

    See Methods for Enzymatic Incorporation of ChromaTide™ dUTPs for specific guidelines for each of these methods.

    Alexa Fluor™ and BODIPY™ Fluorescent Dyes Make Excellent Probes
    Probes made with labeled nucleotides can be used for multicolor techniques such as in situ hybridization and hybridization to arrays. Our proprietary BODIPY™ and Alexa Fluor™ dye conjugates are exceptionally bright, photostable, and essentially pH insensitive. The narrow emission profile of the BODIPY™ dyes helps ensure minimal spectral overlap. The Alexa Fluor™ dyes are highly water soluble, as are DNA probes made from them, making them the labels of choice for fluorescence in situ hybridization.

    Long Linkers Improve Performance
    The ChromaTide™ dUTP and UTP nucleotides are modified at the C-5 position of uridine via a unique alkynylamino linker, which provides a spacer between the nucleotide and the dye to reduce interactions between them. The number in the product name, e.g., the “12” in fluorescein-12-dUTP, indicates the net length of the spacer, in atoms. These spacers result in brighter conjugates and increased hapten accessibility for secondary detection reagents.

    For complete listing of our ChromaTide™ Reagents: Molecular Probes ChromaTide™ and aha labeled nucleotides—Table 8.5.
    For additional information on these labeling reagents, read Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -30°C) and protect from light.
    Labeling Method Direct Labeling
    Shipping Condition Dry Ice
    Concentration 1 mM
    Label or Dye Alexa Fluor™ 568
    Quantity 25 μL
    Product Line Alexa Fluor, ChromaTide
    I'm getting high background after labeling with ChromaTide nucleotides. What do you recommend I do?

    You can try to purify the ChromaTide labeled probe with an appropriate spin column-based method to remove unincorporated ChromaTide nucleotides. Ethanol precipitation may not efficiently remove the unincorporated ChromaTide nucleotides, so a spin column will need to be used.

    The nucleic acid probe is not fluorescent after labeling with ChromaTide nucleotides. What do you recommend I try?

    - Check the base-to-dye ratio to determine the level of incorporation of the ChromaTide nucleotides. Since fluorescent detection may be affected by underlabeling, overlabeling, instrument sensitivity, or other factors, the base-to-dye ratio is a better indicator of incorporation efficiency.
    - ChromaTide nucleotides may not have been incorporated well in the enzymatic labeling reaction. Make sure that the enzymatic method used is compatible with the particular fluorescent ChromaTide nucleotide, since some methods may not be appropriate for all applications. You may also need to further optimize the enzymatic incorporation method, for example by optimizing enzyme concentration, incubation time, concentration, and ratio of labeled and unlabeled nucleotides. For PCR, a lower fidelity polymerase may give higher incorporation rates; however, incorporation rates will be generally low using PCR.
    - Check the fluorescent filter used for detection to make sure it is compatible with the dye. You can also test a small drop of the undiluted fluorescent ChromaTide nucleotide in your filter to make sure you can image the dye alone before it is conjugated to the oligonucleotide. The fluorescence emission of Alexa Fluor 647 is not visible by eye and will require a far-red imaging system for detection.

    Can ChromaTide nucleotides be used for labeling nucleic acids in live cells?

    No, they are not cell permeant so they are only suitable for in vitro incorporation methods. The fluorescent dyes and phosphate groups are too highly charged to allow the nucleotides to penetrate the membrane of an intact cell. Nonfluorescent nucleosides without phosphates such as EdU, EU, or BrdU can be used for live cell nucleic acid incorporation studies.

    How do I determine the incorporation efficiency of the ChromaTide Labeling Nucleotides after enzymatic incorporation?

    The base-to-dye ratio is determined by measuring the absorbance of the nucleic acid at 260 nm and the absorbance of the dye at its absorbance maximum. Using the extinction coefficients for the appropriate dye and nucleic acid, you can then calculate the base-to-dye ratio for the labeled nucleic acid using the Beer-Lambert law. Detailed instructions can be found in these product manuals: (http://tools.thermofisher.com/content/sfs/manuals/td07604.pdf, http://tools.thermofisher.com/content/sfs/manuals/td07605.pdf).

    What is the average dye to base incorporation rate when enzymatically incorporating ChromaTide nucleotides?

    The average incorporation is one dye for every 100-150 bases, so the ChromaTide fluorescently labeled nucleotides typically produce the lowest labeling rates of the nucleic acid labeling methods we offer.

    Can PCR be used to incorporate ChromaTide dUTPs?

    Although we have a protocol for incorporation using PCR, we do not recommend PCR as the best method for incorporation of any of our fluorescent or modified nucleotides. With PCR, incorporation rates are very low. We had some success with Taq polymerases in the past; however, many of the modern DNA polymerases are higher fidelity; as a result, they do not incorporate modified nucleotides very well. This paper below provides a comparison of commercially available polymerases and PCR incorporation efficiency related to type of dye or modification:Anderson JP, Angerer B, Loeb LA (2005) Incorporation of reporter-labeled nucleotides by DNA polymerases. Biotechniques 38:257–264.

    What methods can I use to enzymatically incorporate ChromaTide nucleotides?

    ChromaTide dUTPs are most efficiently incorporated into DNA using nick translation, random primer labeling, end-labeling with terminal deoxynucleotidyl transferase (TdT), and reverse transcription. ChromaTide UTPs can be incorporated into RNA using in vitro transcription. You can find example protocols using these enzymatic reactions below. Specific fluorescent ChromaTide nucleotides may not be appropriate for certain enzymatic methods, so we recommend consulting Table 1 in the ChromaTide product manual prior to purchase. Some ChromaTide products have been discontinued, so not all of the listed ChromaTide nucleotides may be currently available (http://tools.thermofisher.com/content/sfs/manuals/mp07603.pdf, http://tools.thermofisher.com/content/sfs/manuals/td07604.pdf, http://tools.thermofisher.com/content/sfs/manuals/td07605.pdf).

    Where is the fluorescent dye attached on the ChromaTide dUTP and UTP?

    The ChromaTide dUTP and UTP nucleotides are modified at the C-5 position of uridine via a unique alkynylamino linker, which provides a spacer between the nucleotide and the dye to reduce interactions between them. Several of these nucleotides also contain an additional spacer, further separating the label from the nucleic acid. The number in the product name, e.g., the “12” in fluorescein-12-dUTP, indicates the net length of the spacer, in atoms.

    What are ChromaTide Labeling Nucleotides?

    ChromaTide dye-labeled dUTP and UTP nucleotides can be used to synthesize labeled nucleic acid probes without the need for hazardous and expensive radioisotope-labeled nucleotides. You can incorporate these nucleotides using standard molecular biology techniques, and then use the labeled probes for in situ hybridization, microarrays, or blotting. ChromaTide dye-labeled nucleotides are available with different fluorescent colors to facilitate multicolor analysis.

    Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?

    You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.


    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.