Learn More
Active Motif Chromatin Assembly Kit
SDP
Catalog No. NC0198786
Encompass
The Chromatin Assembly Kit enables you to study your DNA sequence of interest in a native chromatin environment. The simple, easy-to-follow protocol generates assembled chromatin in hours with few manipulations, providing you with material that is ideal for downstream applications such as in vitro ChIP, transcription and histone modification assays.
Catalog No. NC0198786 Supplier Active Motif Supplier No. 53500
The Chromatin Assembly Kit enables you to study your DNA sequence of interest in a native chromatin environment. The simple, easy-to-follow protocol generates assembled chromatin in hours with few manipulations, providing you with material that is ideal for downstream applications such as in vitro ChIP, transcription and histone modification assays.