Learn More
Abcam Chromatin E x traction Kit, 100 Tests
Catalog No. NC1659918
Encompass_Preferred
60-minute chromatin extraction kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells and tissues in a simple and rapid format. - Cited in over 50 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1659918 Supplier Abcam Supplier No. AB117152100TEST
60-minute chromatin extraction kit is a complete set of optimized buffers and reagents for isolating chromatin or DNA-protein complex from mammalian cells and tissues in a simple and rapid format. - Cited in over 50 publications
The product is subject to the following: Abcam Restricted Use Statement