Learn More
BUCHI Chromatography Adapter Kits
SDP
Catalog No. 05405491 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Application):
Flash & Prep Chromatography
Flash Chromatography
Prep Chromatography
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
05-405-491 Flash & Prep Chromatography
05-405-489 Flash Chromatography
05-405-490 Prep Chromatography
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-491 Supplier BUCHI Supplier No. 11068363
Add to Cart
Add to Cart
Advanced adapter kits for flash/prep chromatography.
  • For use with BUCHI Pure Chromatography Systems
Specifications
Type Kit
Includes Tube Cutter, Wrenches, Fittings (1/8 in., 1/16 in.), One Piece Fitting (1/16 in.), Tubing (Pneumatic, FEP 1/8 in. and 1/16 in., PEEK 1/16 in.), Stainless Steel Nut/Ferrule, Reductions, Luer Lock Adapters, Fuses, and Injection Valve
For Use With (Application) Flash & Prep Chromatography
For Use With (Equipment) Pure Chromatography Systems