Learn More
Chemglass Life Sciences Column, Chromatography, 35/20 Socket Joint, 1000mL Reservoir, 2 1/2in ID X 18in E.L., 4mm Stpk
SDP
Supplier:  Chemglass Life Sciences CG120234
Encompass_Preferred
Column, Chromatography, 35/20 Socket Joint, 1000mL Reservoir, 2 1/2in ID X 18in E.L., 4mm Stopcock. General purpose column having a PTFE stopcock, reservoir of the listed capacity and a top spherical socket joint. Glass wool (not supplied) is used to support the column packing. Column is constructed using MEDIUM WALL TUBING. Can be used as a gravity eluted column or a flash column (as described by W.C. Still, M. Kahn and A. Mitra of Columbia University in JOURNAL OF ORGANIC CHEMISTRY 43, 2923 [1978], and covered in U.S. Patent Number 4,293,422 dated October 6, 1981) when used with CG-1201 flow control adapter.
Catalog No. CG120234