Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Chromatography of Plant Pigments Kit
GREENER_CHOICESCI_ED

Catalog No. S06970 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06970 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S06970 Supplier Innovating Science™ Supplier No. IS3051
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Students confirm the presence of and learn the roles of different plant pigments using chromatography.

  • Students learn that chlorophyll is necessary for photosynthesis
  • Discover other pigments that are involved in this process
  • Extract various pigments from green plants and separate them with chromatography
  • Learn the roles of these hidden pigments
  • Kit Includes:
    • Chromatography Solvent (250 mL)
    • Plant pigment extraction solvent (Ethanol, 100 mL)
    • 15 microscope slides
    • 15 pieces of chromatography paper
    • 15 capillary tubes

Order Info

For 15 groups

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Confirming the presence and learning the roles of different plant pigments using chromatography
Quantity 1 Kit
Dimensions (L x W x H) 7.25 x 7.125 x 10.5 in.
Class Size 15 Lab Groups
Product Type Chromatography of Plant Pigment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.