Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ ChromatoPur™ Bovine Thrombin, Low Specific Activity, Diagnostic Grade

Catalog No. ICN990780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
100 kU
2 kU
25 kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN990780 100 kU
ICN990725 25 kU
MP219990710 10 kU
ICN990702 2 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN990780 Supplier MP Biomedicals™ Supplier No. 0219990780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable as a diagnostic and research reagent in clotting assays

Chromatographically purified with minimal loss of native clotting activity and minimized degradation to the gamma form. Enzyme activity is measured by clotting assay. 90 to 300U/mg protein.

Physical Characteristics

  • Appearance: White to off-white powder
  • pH: 7.0 to 8.0
  • Protein: ≥50%
  • Specific activity: 90 to 300U/mg protein
  • Activity: Report μ/mg solid
  • Total Activity: U/vial
  • Exipients: Reported
Certifications: ISO, cGMP

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture, Microbial Culture
Name Bovine Thrombin
pH Range 7 to 8
Quantity 100 kU
Source Bovine Thrombin
Storage Requirements -10°C
Form Solid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.