Learn More
Hach Company ChromaVer 3 Chromium Reagent Powder Pillows, 5 or 10 mL, pk/100
Catalog No. 50465355 Shop All Hach Company Products
Encompass
For determination of hexavalent chromium by the Diphenylcarbohydrazide method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L. Kit Sample Size: 5 mL. Kit Range: Up to 1.50 Pack of 100 powder pillows.
Catalog No. 50-465-355 Supplier Hach Company Supplier No. 1271099
For determination of hexavalent chromium by the Diphenylcarbohydrazide method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L. Kit Sample Size: 5 mL. Kit Range: Up to 1.50 Pack of 100 powder pillows.