Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 120 C18 LC Analytical Column, 3 μm, 4.6 x 250 mm, Stainless Steel
Catalog No. 5000332559
Encompass_Preferred
ChromCore 120 C18 columns are based on monodispersed high-purity porous silica particles densely bonded with C18 and exhaustive end-capping for minimal undesired silanol activity They feasure high column efficiency excellent peak shape and good column-to-column reproducibility making them suitable for a broad range of analytes with low to high hydrophobicity
Catalog No. 50-003-32559 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00103001204625S
ChromCore 120 C18 columns are based on monodispersed high-purity porous silica particles densely bonded with C18 and exhaustive end-capping for minimal undesired silanol activity They feasure high column efficiency excellent peak shape and good column-to-column reproducibility making them suitable for a broad range of analytes with low to high hydrophobicity