Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 120 C8 LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Catalog No. 5000332518
Encompass_Preferred
ChromCore 120 C8 columns are based on monodispersed high-purity porous silica particles densely bonded with C8 ligand and exhaustive end-capping to minimize undesired silanol activity designed for separating analytes with intermediate to high hydrophobicity
Catalog No. 50-003-32518 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00705001204625S
ChromCore 120 C8 columns are based on monodispersed high-purity porous silica particles densely bonded with C8 ligand and exhaustive end-capping to minimize undesired silanol activity designed for separating analytes with intermediate to high hydrophobicity