Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 120 C8 LC Analytical Column, 3 μm, 3.0 x 100 mm, Stainless Steel
Catalog No. 5000332587
Encompass_Preferred
ChromCore 120 C8 columns are based on monodispersed high-purity porous silica particles densely bonded with C8 ligand and exhaustive end-capping to minimize undesired silanol activity designed for separating analytes with intermediate to high hydrophobicity
Catalog No. 50-003-32587 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00703001203010S
ChromCore 120 C8 columns are based on monodispersed high-purity porous silica particles densely bonded with C8 ligand and exhaustive end-capping to minimize undesired silanol activity designed for separating analytes with intermediate to high hydrophobicity