Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 300 C18 LC Analytical Column, 3 μm, 2.1 x 100 mm, Stainless Steel
Catalog No. 5000332543
Encompass_Preferred
ChromCore 300 C18 columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C18 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins
Catalog No. 50-003-32543 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00103003002110S
Add to Cart
Add to Cart
ChromCore 300 C18 columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C18 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins