Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 300 C4-T LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Catalog No. 5000332565
Encompass_Preferred
ChromCore 300 C4-T columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C4 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins
Catalog No. 50-003-32565 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A22603003004610S
ChromCore 300 C4-T columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C4 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins