Learn More
NANOMICRO TECHNOLOGIES INC ChromCore 300 C8 LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Catalog No. 5000332590
Encompass_Preferred
ChromCore 300 C8 columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C8 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins
Catalog No. 50-003-32590 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00703003004610S
Add to Cart
Add to Cart
ChromCore 300 C8 columns are based on monodispersed 300 pore high-purity spherical silica particles modified with C8 ligand and exhaustive end-capping They are designed for separating analytes of large size such as ginsenosides PEG derivatives peptides and small proteins