Learn More
NANOMICRO TECHNOLOGIES INC ChromCore AQ C18 LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Catalog No. 5000332576
Encompass_Preferred
ChromCore AQ C18 columns are based on proprietary C18 modified monodispersed silica particles for excellent aqueous compatibility They are the column of choice for applications requiring highly aqueous mobile phase and/or C18 selectivity
Catalog No. 50-003-32576 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A20103001804610S
ChromCore AQ C18 columns are based on proprietary C18 modified monodispersed silica particles for excellent aqueous compatibility They are the column of choice for applications requiring highly aqueous mobile phase and/or C18 selectivity