Learn More
NANOMICRO TECHNOLOGIES INC ChromCore AR C18 LC Analytical Column, 5 μm, 4.6 x 150 mm, Stainless Steel
Catalog No. 5000332569
Encompass_Preferred
ChromCore AR C18 columns are based on bonding a unique sterically protected C18 silane to the surface of high-purity monodispersed porous silica particles designed for applications that require extreme acidic conditions highly aqueous mobile phases and/or selectivity complimentary to ChromCore C18 columns
Catalog No. 50-003-32569 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A40105001204615S
ChromCore AR C18 columns are based on bonding a unique sterically protected C18 silane to the surface of high-purity monodispersed porous silica particles designed for applications that require extreme acidic conditions highly aqueous mobile phases and/or selectivity complimentary to ChromCore C18 columns