Learn More
NANOMICRO TECHNOLOGIES INC ChromCore HILIC-Diol LC Analytical Column, 3 μm, 4.6 x 150 mm, Stainless Steel
Catalog No. 5000332560
Encompass_Preferred
ChromCore HILIC-Diol columns are based on proprietary diol modified monodispersed high-purity silica particles They provide a neutral surface with intermediate polarity suited for separating pesticides herbicides pharmaceutical metabolites and polar natural products
Catalog No. 50-003-32560 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A02003001204615S
ChromCore HILIC-Diol columns are based on proprietary diol modified monodispersed high-purity silica particles They provide a neutral surface with intermediate polarity suited for separating pesticides herbicides pharmaceutical metabolites and polar natural products