Learn More
NANOMICRO TECHNOLOGIES INC ChromCore HILIC-Diol LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Catalog No. 5000332585
Encompass_Preferred
ChromCore HILIC-Diol columns are based on proprietary diol modified monodispersed high-purity silica particles They provide a neutral surface with intermediate polarity suited for separating pesticides herbicides pharmaceutical metabolites and polar natural products
Catalog No. 50-003-32585 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A02005001204625S
ChromCore HILIC-Diol columns are based on proprietary diol modified monodispersed high-purity silica particles They provide a neutral surface with intermediate polarity suited for separating pesticides herbicides pharmaceutical metabolites and polar natural products