Learn More
NANOMICRO TECHNOLOGIES INC ChromCore NH2 LC Analytical Column, 3 μm, 4.6 x 150 mm, Stainless Steel
Catalog No. 5000332555
Encompass_Preferred
ChromCore NH2 columns are based on monodispersed high-purity porous silica particles bonded with propylamino ligand designed for separating polar and hydrophilic analytes such as monosaccharides oligosaccharides and sugar alcohols under HILIC conditions
Catalog No. 50-003-32555 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00803001204615S
ChromCore NH2 columns are based on monodispersed high-purity porous silica particles bonded with propylamino ligand designed for separating polar and hydrophilic analytes such as monosaccharides oligosaccharides and sugar alcohols under HILIC conditions