Learn More
NANOMICRO TECHNOLOGIES INC ChromCore NH2 LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Catalog No. 5000332580
Encompass_Preferred
ChromCore NH2 columns are based on monodispersed high-purity porous silica particles bonded with propylamino ligand designed for separating polar and hydrophilic analytes such as monosaccharides oligosaccharides and sugar alcohols under HILIC conditions
Catalog No. 50-003-32580 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00805001204625S
ChromCore NH2 columns are based on monodispersed high-purity porous silica particles bonded with propylamino ligand designed for separating polar and hydrophilic analytes such as monosaccharides oligosaccharides and sugar alcohols under HILIC conditions