Learn More
NANOMICRO TECHNOLOGIES INC ChromCore PFP LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Catalog No. 5000332586
Encompass_Preferred
ChromCore PFP columns are based on monodispersed high-purity porous silica particles bonded with pentafluorobenzene (PFP) ligand and exhaustive end-capping They are designed for a broad range of applications that require selectivity different from ChromCore Phenyl for aromatic analytes such as halogenated aromatic compounds and Vitamin E isomers
Catalog No. 50-003-32586 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A04303001204610S
ChromCore PFP columns are based on monodispersed high-purity porous silica particles bonded with pentafluorobenzene (PFP) ligand and exhaustive end-capping They are designed for a broad range of applications that require selectivity different from ChromCore Phenyl for aromatic analytes such as halogenated aromatic compounds and Vitamin E isomers