Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Phenyl-Hexyl LC Analytical Column, 3 μm, 4.6 x 100 mm, Stainless Steel
Catalog No. 5000332593
Encompass_Preferred
ChromCore Phenyl-Hexyl columns are based on monodispersed high-purity porous silica particles bonded with hexylbenzene ligand and exhaustive end-capping They are designed for a broad range of applications that require enhanced hydrophobic retention and selectivity towards aromatic analytes such as isomers of aromatic and heterocyclic pharmaceuticals and catechins
Catalog No. 50-003-32593 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A31103001204610S
ChromCore Phenyl-Hexyl columns are based on monodispersed high-purity porous silica particles bonded with hexylbenzene ligand and exhaustive end-capping They are designed for a broad range of applications that require enhanced hydrophobic retention and selectivity towards aromatic analytes such as isomers of aromatic and heterocyclic pharmaceuticals and catechins