Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Phenyl LC Analytical Column, 3 μm, 4.6 x 150 mm, Stainless Steel
Catalog No. 5000332581
Encompass_Preferred
ChromCore Phenyl columns are based on monodispersed high-purity porous silica particles bonded with propylbenzene ligand and exhaustive end-capping They are designed for a broad range of applications that require selectivity towards aromatic analytes such as isomers of aromatic and heterocyclic pharmaceuticals and catechins
Catalog No. 50-003-32581 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A01103001204615S
ChromCore Phenyl columns are based on monodispersed high-purity porous silica particles bonded with propylbenzene ligand and exhaustive end-capping They are designed for a broad range of applications that require selectivity towards aromatic analytes such as isomers of aromatic and heterocyclic pharmaceuticals and catechins