Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Polar C18 LC Analytical Column, 3 μm, 3.0 x 150 mm, Stainless Steel
Catalog No. 5000332552
Encompass_Preferred
ChromCore Polar C18 columns are based on amide-embedded C18 modified monodispersed porous silica particles with exhaustive end-capping designed for applications that require highly aqueous mobile phase and/or selectivity different from conventional C18 columns
Catalog No. 50-003-32552 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A06003001203015S
ChromCore Polar C18 columns are based on amide-embedded C18 modified monodispersed porous silica particles with exhaustive end-capping designed for applications that require highly aqueous mobile phase and/or selectivity different from conventional C18 columns