Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Silica LC Analytical Column, 3 μm, 4.6 x 150 mm, Stainless Steel
Catalog No. 5000332522
Encompass_Preferred
ChromCore Silica columns are based on unmodified monodispersed high-purity porous silica particles designed for separating highly hydrophobic compounds under normal phase conditions
Catalog No. 50-003-32522 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00303001204615S
Add to Cart
Add to Cart
ChromCore Silica columns are based on unmodified monodispersed high-purity porous silica particles designed for separating highly hydrophobic compounds under normal phase conditions