Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Silica LC Analytical Column, 5 μm, 4.6 x 250 mm, Stainless Steel
Catalog No. 5000332579
Encompass_Preferred
ChromCore Silica columns are based on unmodified monodispersed high-purity porous silica particles designed for separating highly hydrophobic compounds under normal phase conditions
Catalog No. 50-003-32579 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A00305001204625S
ChromCore Silica columns are based on unmodified monodispersed high-purity porous silica particles designed for separating highly hydrophobic compounds under normal phase conditions