Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Sugar-10Ca LC Analytical Column, 6 μm, 7.8 x 300 mm, Stainless Steel
Catalog No. 5000332519
Encompass_Preferred
ChromCore Suga-10Ca columns are based on sulfonated monodispersed spherical PS/DVB particles loaded with Ca2 ions designed for separating sugars and sugar alcohols based on ligand exchange and size exclusion mechanism
Catalog No. 50-003-32519 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A01906001007830S
ChromCore Suga-10Ca columns are based on sulfonated monodispersed spherical PS/DVB particles loaded with Ca2 ions designed for separating sugars and sugar alcohols based on ligand exchange and size exclusion mechanism