Learn More
NANOMICRO TECHNOLOGIES INC ChromCore Sugar-10H LC Analytical Column, 6 μm, 7.8 x 300 mm, Stainless Steel
Catalog No. 5000332508
Encompass_Preferred
ChromCore Suga-10Ca columns are based on sulfonated monodispersed spherical PS/DVB particles in the hydronium form generally used to separate organic acids and sugars mainly based on ion exclusion and size exclusion mechanism
Catalog No. 50-003-32508 Supplier NANOMICRO TECHNOLOGIES INC Supplier No. A01706001007830S
Add to Cart
Add to Cart
ChromCore Suga-10Ca columns are based on sulfonated monodispersed spherical PS/DVB particles in the hydronium form generally used to separate organic acids and sugars mainly based on ion exclusion and size exclusion mechanism