Learn More
Newcomer Supply Chrome Alum-Gelatin Adhesive, 250 ml
SDP
Catalog No. NC1692262
Encompass
For water bath use to add extra adhesion of the tissue section to the glass slide. Newcomer Supply Chrome Alum-Gelatin Adhesive provides a blended solution of chrome alum and high-quality gelatin that promotes a strong adhesive bond between tissue sections and microscopic slides. Chrome Alum-Gelatin Adhesive can be used as an additive to water baths or as subbed slide/direct slide coating application to prevent or reduce the loss of tissue sections due to the nature of the tissue, section thickness or harsh staining treatments, while leaving minimal or no background staining. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1692262 Supplier Newcomer Supply Supplier No. 1033A250ML
For water bath use to add extra adhesion of the tissue section to the glass slide. Newcomer Supply Chrome Alum-Gelatin Adhesive provides a blended solution of chrome alum and high-quality gelatin that promotes a strong adhesive bond between tissue sections and microscopic slides. Chrome Alum-Gelatin Adhesive can be used as an additive to water baths or as subbed slide/direct slide coating application to prevent or reduce the loss of tissue sections due to the nature of the tissue, section thickness or harsh staining treatments, while leaving minimal or no background staining. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.