Learn More
MSC Proto Combination Wrench, 13/16 in, 12 Point, 1 ea
Catalog No. 501920024
Encompass
The Proto J1226-T500 combination wrench is engineered for demanding applications, featuring a durable alloy steel construction and a full polished chrome finish. It is designed to provide excellent grip and torque transfer, making it suitable for professional use in various industrial and mechanical settings.
  • Full polish finish provides corrosion resistance and easy-to-clean surfaces
  • A 12 point box end allows for fine indexing, helping to improve speed and access
  • Fits 4, 6 and 12 point fasteners
  • Anti-slip design (ASD) open end
  • Box ends incorporate the TorquePlus system
  • Reduces risk of fastener rounding, provides a better grip on worn, rounded fasteners
  • Allows up to 15% more torque to be applied to the fastener
Catalog No. 50-192-0024 Supplier MSC Supplier No. 84974252
The Proto J1226-T500 combination wrench is engineered for demanding applications, featuring a durable alloy steel construction and a full polished chrome finish. It is designed to provide excellent grip and torque transfer, making it suitable for professional use in various industrial and mechanical settings.
  • Full polish finish provides corrosion resistance and easy-to-clean surfaces
  • A 12 point box end allows for fine indexing, helping to improve speed and access
  • Fits 4, 6 and 12 point fasteners
  • Anti-slip design (ASD) open end
  • Box ends incorporate the TorquePlus system
  • Reduces risk of fastener rounding, provides a better grip on worn, rounded fasteners
  • Allows up to 15% more torque to be applied to the fastener