Learn More
Supertek™ Chrome Plated Steel Set of Tuning Forks
GREENER_CHOICESCI_ED
Catalog No. S81868 Shop All Supertek Scientific Products
Change view
Click to view available options
Quantity:
13 Tuning Forks/Set
8 Tuning Forks/Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81868 13 Tuning Forks/Set
S81867 8 Tuning Forks/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S81868 Supplier Supertek™ Supplier No. PH36129/3
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Supertek™ Chrome Plated Steel Set of Tuning Forks is an economical set. Comes in a Styrofoam case.
  • Made of steel
  • Chromatic scale
  • Frequency 256 to 512
  • In Styrofoam case
  • Chrome Plated
Specifications
Product Type Tuning Fork
Material Chrome-plated Steel
Includes 13 Tuning Forks, Styrofoam Case
Quantity 13 Tuning Forks/Set
Frequency 100, 128, 256, 288, 320, 341.3, 384, 426.6, 480, 512, 1024, 2048, 4096 Hz