Learn More
Medchemexpress LLC Chromeceptin 10mg | 331859-86-0 | 10 MG
SDP
Catalog No. 5000227447
Encompass_Preferred
Chromeceptin is a small-molecule IGF signaling pathway inhibitor for research use that suppresses IGF2 expression and reduces phosphorylation of AKT and mTOR. Supplied as a high-purity solid suitable for biochemical and cell-based studies.
  • High purity suitable for biochemical assays and cell-based studies.
  • Inhibits IGF signaling and suppresses IGF2 expression.
  • Reduces phosphorylation of AKT and mTOR in target cells.
  • Available as a 10 mg powder with recommended low-temperature storage.
  • Soluble in common organic solvents for assay preparation.
Catalog No. 50-002-27447 Supplier Medchemexpress LLC Supplier No. HY11544910MG
Chromeceptin is a small-molecule IGF signaling pathway inhibitor for research use that suppresses IGF2 expression and reduces phosphorylation of AKT and mTOR. Supplied as a high-purity solid suitable for biochemical and cell-based studies.
  • High purity suitable for biochemical assays and cell-based studies.
  • Inhibits IGF signaling and suppresses IGF2 expression.
  • Reduces phosphorylation of AKT and mTOR in target cells.
  • Available as a 10 mg powder with recommended low-temperature storage.
  • Soluble in common organic solvents for assay preparation.