Learn More
TARGETMOL CHEMICALS INC CHROMECEPTIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T600555MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Chromeceptin is an inhibitor of the IGF signaling pathway decreases the numbers of tumorsphere and inhibits the AKT/mTOR pathway. purity: 99%
Catalog No. 50-257-9687