Learn More
Medchemexpress LLC Chromeceptin 5mg | 331859-86-0 | 359.35 g·mol⁻1 | C19H16F3N3O | 5 MG
SDP
Catalog No. 5000229950
Encompass_Preferred
Chromeceptin is a small-molecule research inhibitor of the IGF signaling pathway that suppresses IGF2 expression and reduces phosphorylation of AKT and mTOR in hepatocyte and hepatocellular carcinoma cells. Supplied for preclinical laboratory use.
  • IGF signaling pathway inhibitor.
  • Suppresses IGF2 expression at mRNA and protein levels.
  • Reduces phosphorylation of AKT and mTOR.
  • High chemical purity suitable for preclinical research.
  • Available in milligram pack sizes and DMSO solution formats for convenient dosing.
Catalog No. 50-002-29950 Supplier Medchemexpress LLC Supplier No. HY1154495MG
Chromeceptin is a small-molecule research inhibitor of the IGF signaling pathway that suppresses IGF2 expression and reduces phosphorylation of AKT and mTOR in hepatocyte and hepatocellular carcinoma cells. Supplied for preclinical laboratory use.
  • IGF signaling pathway inhibitor.
  • Suppresses IGF2 expression at mRNA and protein levels.
  • Reduces phosphorylation of AKT and mTOR.
  • High chemical purity suitable for preclinical research.
  • Available in milligram pack sizes and DMSO solution formats for convenient dosing.