Learn More
Sigma Aldrich Fine Chemicals Biosciences Chromeo P503, 1 mg
Catalog No. NC1808882
Encompass
Chromeo P503 is used as a fluorogenic reagent to label primary amine groups within molecules such as proteins. Chromeo P503 labeled proteins may be studied in a variety of gel electrophoresis and chromatographic applications.
Catalog No. NC1808882 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 306931MG
Chromeo P503 is used as a fluorogenic reagent to label primary amine groups within molecules such as proteins. Chromeo P503 labeled proteins may be studied in a variety of gel electrophoresis and chromatographic applications.