Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chromic Acid, Polymer-Supported

Catalog No. 35982325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-982-325G 25 g
35-982-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 35-982-325G Supplier Sigma Aldrich Supplier No. 35982325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polymer-supported reagent for the clean oxidation of primary and secondary alcohols to carbonyl compounds in high yields.Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Specifications

MDL Number MFCD00010952
Quantity 25 g
Synonym Chromic acid on Amberlyst(R) A-26
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.