Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Chloride, 98%, Spectrum™ Chemical
SDP

Catalog No. 18609743 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-743 500 g Amber Glass Bottle
18-609-741 25 g Amber Glass Bottle
18-609-739 125 g Amber Glass Bottle
18-609-738 2.5 kg Poly Pail
18-609-740 12 kg Poly Pail
18-609-742 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-609-743 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1238500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10060-12-5

Specifications

CAS Max % 1
Physical Form Crystals
Quantity 500 g
Molecular Weight (g/mol) 266.45
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Chromic Chloride Hexahydrate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.